Learn More
Phenytoin sodium, MedChemExpress
Product Code. 16472179
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16472179
Brand: Medchem Express HYB0448A
MedChemExpress Phenytoin sodium (5,5-Diphenylhydantoin sodium salt) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.
Phenytoin sodium (5,5-Diphenylhydantoin sodium salt) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice.
Specifications
Chemical Name or Material Phenytoin sodium
Physical Form Solid
CAS 630-93-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C15H11N2NaO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 5,5-Diphenylhydantoin sodium salt
Solubility Information DMSO : 50 mg/mL (182.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.65 mM; Need ultrasonic)
SMILES O=C1N=C(O[Na])NC1(C2=CC=CC=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 274.25
Formula Weight 274.25
Percent Purity 99.96%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.