Learn More
Phenyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals
Catalog Number p-184266
376.88 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4207-56-1
C9H14Br3N
375.92
MFCD00011789
RHXJPCQWYPXLEH-UHFFFAOYSA-N
mono n,n,n-trimethylbenzenaminium tribromide
21525000
trimethyl(phenyl)azanium;triiodide
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4207-56-1
C9H14Br3N
375.92
MFCD00011789
RHXJPCQWYPXLEH-UHFFFAOYSA-N
mono n,n,n-trimethylbenzenaminium tribromide
21525000
trimethyl(phenyl)azanium;triiodide
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Melting Point 113.0°C to 117.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula C6H5N(CH3)3Br·Br2
Quantity 25 g
Fieser 01,855; 02,328; 04,386
Solubility Information Solubility in water: insoluble.
Formula Weight 375.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenyltrimethylammonium tribromide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Phenyltrimethylammonium tribromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.