missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylselenenyl bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.
Solubility
Soluble in toluene, very faint turbidity.
Notes
Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Employed in the synthesis of enones by selenenylation of α-cyano ketones with lithium naphthalenide. Phenylselenyl bromide was used in the preparation of exo-2-methyl-7-phenylselenyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione and endo-2-methyl-7-phenylselanyl-octahydro-8-oxa-2,7b-diaza-dicyclopenta[a,e]pentalene-1,3-dione.
Solubility
Soluble in toluene, very faint turbidity.
Notes
Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 56°C to 62°C |
Boiling Point | 107°C to 108°C (15 mmHg) |
Quantity | 1 g |
UN Number | UN2928 |
Beilstein | 2039772 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in toluene,very faint turbidity. |
Formula Weight | 235.98 |
Percent Purity | 98% |
Chemical Name or Material | Phenylselenenyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'