missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylazoformic acid 2-phenylhydrazide, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is a reagent for mercury determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a reagent for mercury determination.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is a reagent for mercury determination.
Solubility
Soluble in acetone. Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 119°C to 123°C |
Quantity | 5 g |
Beilstein | 959475 |
Solubility Information | Soluble in acetone. Insoluble in water. |
Formula Weight | 240.26 |
Chemical Name or Material | Phenylazoformic acid 2-phenylhydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'