missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoromethyl sulfide, 98%, Thermo Scientific Chemicals

Catalog Number p-7045277
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
456-56-4
C7H5F3S
178.17
MFCD00040839
YQQKTCBMKQQOSM-UHFFFAOYSA-N
phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
68011
trifluoromethylsulfanylbenzene
FC(F)(F)SC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
456-56-4
C7H5F3S
178.17
MFCD00040839
YQQKTCBMKQQOSM-UHFFFAOYSA-N
phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
68011
trifluoromethylsulfanylbenzene
FC(F)(F)SC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.249
Boiling Point 140°C to 142°C
Flash Point 37°C (98°F)
Odor Unpleasant
Refractive Index 1.4655
Quantity 1 g
UN Number UN1993
Beilstein 2043134
Formula Weight 178.18
Percent Purity 98%
Chemical Name or Material Phenyl trifluoromethyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'