Learn More
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Catalog Number p-7025286
320.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
118-55-8
C13H10O3
214.22
MFCD00002213
ZQBAKBUEJOMQEX-UHFFFAOYSA-N
phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
8361
CHEBI:34918
phenyl 2-hydroxybenzoate
C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
118-55-8
C13H10O3
214.22
MFCD00002213
ZQBAKBUEJOMQEX-UHFFFAOYSA-N
phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
8361
CHEBI:34918
phenyl 2-hydroxybenzoate
C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY
Melting Point 41°C to 45°C
Boiling Point 172°C to 174°C (12 mmHg)
Flash Point 160°C (320°F)
Quantity 100 g
Beilstein 393969
Merck Index 14,7310
Solubility Information Soluble in alcohol,ether,chloroform,turpentine,acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Phenyl salicylate