Learn More
Phenol-3,5-d2, CDN
Product Code. 30315512
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315512
Brand: CDN CDND28821G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenol-3,5-D2
CAS 64045-87-0
Molecular Formula C6 D2 H4 O
Quantity 1 g
Synonym Phen-3,5-d2-ol (9 CI, ACI), 3,5-Dideuteriophenol, Phenol-3,5-D2
InChI Formula InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i2D,3 D
SMILES [2 H]c1cc([2 H])cc(O)c1
IUPAC Name 3,5-dideuteriophenol
Molecular Weight (g/mol) 96.124
Formula Weight 96.054 g/mol
Percent Purity 97 atom % D, min 98% Chemical Purity