Learn More
Fisherbrand™ pH Indicator Paper Strips
Provides optimal quality and reliable readings. Fisherbrand™ pH Indicator Paper Strips are supplied in plastic reels that ensure long-term stability and protection against many external influences.
Brand: Fisherbrand
Additional Details : Weight : 0.30000kg
Description
- Ready to use
- Manufactured from high quality filter paper
- Colors of the scales are specially mixed to perfectly match the reaction color of the indicator papers
Specifications
ISO 9001 | |
Roll | |
pH Paper |
6.4 to 8 | |
1 Each |