Learn More
Uvex Perforated Micro Black/Orange PU/RU 65211 Low Shoe - Width 10
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine the limits for safety shoes; range in protection classes S3 and S1 P
1665.00 NOK
Specifications
Color | Black, Orange |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane rubber (PU/RU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17281051
|
Uvex
6521135 |
35 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17291051
|
Uvex
6521136 |
36 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17201061
|
Uvex
6521137 |
37 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17211061
|
Uvex
6521138 |
38 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17221061
|
Uvex
6521139 |
39 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17231061
|
Uvex
6521140 |
40 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17241061
|
Uvex
6521141 |
41 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17251061
|
Uvex
6521142 |
42 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17261061
|
Uvex
6521143 |
43 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17271061
|
Uvex
6521144 |
44 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17281061
|
Uvex
6521145 |
45 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17291061
|
Uvex
6521146 |
46 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17201071
|
Uvex
6521147 |
47 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17211071
|
Uvex
6521148 |
48 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17221071
|
Uvex
6521149 |
49 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17231071
|
Uvex
6521150 |
50 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17241071
|
Uvex
6521151 |
51 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17251071
|
Uvex
6521152 |
52 |
1665.00 NOK
1 pair |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 VIBRAM Perforated Shoe 6521.1 S1 P HRO SRC
- Sporty, multifunctional safety sandal
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-repellent perforated high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Reflecting elements for more safety
- Individually adjustable due to elastic laces with quick-lock mechanism
- Normal laces also provided
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue, dust tongue
- Injection-molded polyurethane scuffcap to protect upper leather on toe cap and significantly extend resistance time
- Meets ESD requirements: resistance to ground Kleiner
- Lightweight two-layer insole with excellent shock absorption characteristics and outstanding slip resistance
- Multilayer sole system comprising PU midsole and highly slip resistant rubber outersole from VIBRAM
- Optimized ankle protection to ensure safety when working on ladders
- Rugged, self-cleaning sole ridges ensure max grip for demanding applications on uneven or dirty floors (SRC labeling)
- Withstands brief exposure to heat approx. 300°C
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6521.1
- Standard EN ISO 20345:2011 S1 P HRO SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane rubber (PU/RU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S1 |