Learn More
Alfa Aesar™ Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%
Brand: Alfa Aesar™ L16614.06
Additional Details : CAS Number : 27639-98-1 Weight : 0.00500kg Transport : UN number : 3265 Chem class : 8 Pack group : II
Quantity | 5g |
---|
Chemical Identifiers
27639-98-1 | |
664.09 | |
IIYQDKPSLAINNX-UHFFFAOYSA-N | |
119616 | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)F |
C12F24O4 | |
MFCD00084743 | |
perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propoxy, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, propanoyl fluoride,2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, acmc-20ajy8, 3,6,9-trioxadodecanoyl fluoride, tetradecafluoro-2,5,8-tris trifluoromethyl | |
2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propanoyl fluoride |
Specifications
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride | |
5g | |
C12F24O4 | |
UN3265 | |
IIYQDKPSLAINNX-UHFFFAOYSA-N | |
2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propanoyl fluoride | |
119616 | |
97% |
158°C to 161°C | |
27639-98-1 | |
MFCD00084743 | |
perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propoxy, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, propanoyl fluoride,2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy, acmc-20ajy8, 3,6,9-trioxadodecanoyl fluoride, tetradecafluoro-2,5,8-tris trifluoromethyl | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)F | |
664.09 | |
664.09 |