missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl isothiocyanate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Density | 1.56 |
Boiling Point | 109°C to 110°C (35 mmHg) |
Refractive Index | 1.5378 |
Quantity | 1 g |
UN Number | UN2922 |
Beilstein | 2736207 |
Sensitivity | Moisture sensitive |
Formula Weight | 225.14 |
Percent Purity | 96% |
Chemical Name or Material | Pentafluorophenyl isothiocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'