missing translation for 'onlineSavingsMsg'
Learn More

Pentachloropyridine, 98%

Product Code. 11481488
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11481488

Brand: Thermo Scientific Alfa Aesar A13842.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2176-62-7 | C5Cl5N | 251.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2176-62-7
Molecular Formula C5Cl5N
Molecular Weight (g/mol) 251.312
MDL Number MFCD00006230
InChI Key DNDPLEAVNVOOQZ-UHFFFAOYSA-N
Synonym pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
PubChem CID 16584
IUPAC Name 2,3,4,5,6-pentachloropyridine
SMILES C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 123°C to 127°C
Quantity 100 g
UN Number UN3077
Beilstein 155197
Formula Weight 251.33
Percent Purity 98%
Chemical Name or Material Pentachloropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.