missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta Antibody (rinucumab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28592JF635
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PDGF R beta | |
| Monoclonal | |
| Janelia Fluor 635 | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| PDGFRB/ CD140b | |
| 0.1 mL | |
| Oncogenes, Phospho Specific, Protein Kinase | |
| 5159 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| rinucumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction