missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PCK1 (Human) Recombinant Protein

Human PCK1 full-length ORF ( AAH23978, 1 a.a. - 622 a.a.) recombinant protein with GST-tag at N-terminal.

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH23978
Gene ID (Entrez) 5105
Name phosphoenolpyruvate carboxykinase 1 (soluble)
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16188771
View Documents Promotion Details
Abnova™
H00005105-P01.10UG
10 μg
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16198771
View Documents Promotion Details
Abnova™
H00005105-P01.25UG
25 μg
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MPPQLQNGLNLSAKVVQGSLDSLPQAVREFLENNAELCQPDHIHICDGSEEENGRLLGQMEEEGILRRLKKYDNCWLALTDPRDVARIESKTVIVTQEQRDTVPIPKTGLSQLGRWMSEEDFEKAFNARFPGCMKGRTMYVIPFSMGPLGSPLSKIGIELTDSPYVVASMRIMTRMGTPVLEALGDGEFVKCLHSVGCPLPLQKPLVNNWPCNPELTLIAHLPDRREIISFGSGYGGNSLLGKKCFALRMASRLAKEEGWLAEHMLVLGITNPEGEKKYLAAAFPSACGKTNLAMMNPSLPGWKVECVGDDIAWMKFDAQGHLRAINPENGFFGVAPGTSVKTNPNAIKTIQKNTIFTNVAETSDGGVYWEGIDEPLASGVTITSWKNKEWSSEDGEPCAHPNSRFCTPASQCPIIDAAWESPEGVPIEGIIFGGRRPAGVPLVYEALSWQHGVFVGAAMRSEATAAAEHKGKIIMHDPFAMRPFFGYNFGKYLAHWLSMAQHPAAKLPKIFHVNWFRKDKEGKFLWPGFGENSRVLEWMFNRIDGKASTKLTPIGYIPKEDALNLKGLGHINMMELFSISKEFWEKEVEDIEKYLEDQVNADLPCEIEREILALKQRISQM
Specifications

Specifications

AAH23978
phosphoenolpyruvate carboxykinase 1 (soluble)
125% SDS-PAGE Stained with Coomassie Blue
MGC22652, PEPCK-C, PEPCK1, PEPCKC
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
5105
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCK1
GST
Videos
SDS
Documents

Documents

Product Certifications