missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCF11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | PCF11 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802385
|
Proteintech
23540-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16892375
|
Proteintech
23540-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to the 173 kDa human PCF11 protein.
Component of pre-mRNA cleavage complex II.Specifications
PCF11 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O94913 | |
51585 | |
PCF11 Fusion Protein Ag20263 | |
Primary | |
-20°C | |
PCF11 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0824, PCF11 | |
PCF11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |