Learn More
Abnova™ PBX1 (Human) Recombinant Protein
Human PBX1 partial ORF (NP_002576.1, 311 a.a. - 403 a.a.) recombinant protein with GST tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002576.1 |
---|---|
Gene ID (Entrez) | 5087 |
Name | pre-B-cell leukemia homeobox 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014505
|
Abnova™
H00005087-Q02.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024505
|
Abnova™
H00005087-Q02.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: VTATNVSAHGSQANSPSTPNSAGSSSSFNMSNSGDLFMSVQSLNGDSYQGAQVGANVQSQVDTLRHVISQTGGYSDGLAASQMYSPQGISANG
Specifications
NP_002576.1 | |
pre-B-cell leukemia homeobox 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
DKFZp686B09108, MGC126627 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5087 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PBX1 | |
GST |