missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARVB Antibody (PARVB/8214R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20827JF585
This item is not returnable.
View return policy
Description
PARVB Monoclonal antibody specifically detects PARVB in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PARVB | |
| Monoclonal | |
| Janelia Fluor 585 | |
| affixin, beta-parvin, CGI-56, parvin, beta | |
| N-terminal region (residues 1-91) of human PARVB | |
| 0.1 mL | |
| Cancer | |
| 29780 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PARVB/8214R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction