missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PAM (Human) Recombinant Protein
Human PAM full-length ORF ( AAH18127, 1 a.a. - 866 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005066-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAGRVPSLLVLLVFPSSCLAFRSPLSVFKRFKETTRPFSNECLGTTRPVVPIDSSDFALDIRMPGVTPKQSDTYFCMSMRIPVDEEAFVIDFKPRASMDTVHHMLLFGCNMPSSTGSYWFCDEGTCTDKANILYAWARNAPPTRLPKGVGFRVGGETGSKYFVLQVHYGDISAFRDNNKDCSGVSLHLTRLPQPLIAGMYLMMSVDTVIPAGEKVVNSDISCHYKNYPMHVFAYRVHTHHLGKVVSGYRVRNGQWTLIGRQSPQLPQAFYPVGHPVDVSFGDLLAARCVFTGEGRTEATHIGGTSSDEMCNLYIMYYMEAKHAVSFMTCTQNVAPDMFRTIPPEANIPIPVKSDMVMMHEHHKETEYKDKIPLLQQPKREEEEVLDQDFHMEEALDWPGVYLLPGQVSGVALDPKNNLVIFHRGDHVWDGNSFDSKFVYQQIGLGPIEEDTILVIDPNNAAVLQSSGKNLFYLPHGLSIDKDGNYWVTDVALHQVFKLDPNNKEGPVLILGRSMQPGSDQNHFCQPTDVAVDPGTGAIYVSDGYCNSRIVQFSPSGKFITQWGEESSGSSPLPGQFTVPHSLALVPLLGQLCVADRENGRIQCFKTDTKEFVREIKHSSFGRNVFAISYIPGLLFAVNGKPHFGDQEPVQGFVMNFSNGEIIDIFKPVRKHFDMPHDIVASEDGTVYIGDAHTNTVWKFTLTEKLEHRSVKKAGIEVQEIKEAEAVVETKMENKPTSSELQKMQEKQKLIKEPGSGVPVVLITTLLVIPVVVLLAIAIFIRWKKSRAFGDSEHKLETSSGRVLGRFRGKGSGGLNLGNFFASRKGYSRKGFDRLSTEGSDQEKEDDGSESEEEYSAPLPALAPSSS
Specifications
AAH18127 | |
peptidylglycine alpha-amidating monooxygenase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAM | |
GST |
5066 | |
Wheat germ expression system | |
25 μg | |
PAL, PHM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |