Learn More
Alfa Aesar™ Palladium wire, 1.0mm (0.04 in.) dia., 99.98+% (metals basis)
Brand: Alfa Aesar™ 040212.CB
Additional Details : CAS Number : 7440-05-3 Weight : 0.01000kg
Quantity | 250cm |
---|
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Palladium wire | |
250cm | |
7440-05-3 | |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
Odorless | |
23938 | |
Wire |
≈9.44g/m | |
1.0mm (0.04 in.) dia. | |
(metals basis) | |
MFCD00011167 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
106.42 | |
CHEBI:33363 | |
≥99.98% |