Learn More
p-Xylylenediamine, 97%, Thermo Scientific Chemicals
417.35 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
50 g
Description
p-Xylylenediamine is
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylylenediamine is
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
p-Xylylenediamine is
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
Specifications
Melting Point | 59°C to 63°C |
Boiling Point | 230°C (10mmHg) |
Flash Point | 134°C (273°F) |
Quantity | 2 g |
UN Number | UN3259 |
Beilstein | 2206190 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 136.2 |
Percent Purity | 97% |