missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
p-Tolylhydrazine hydrochloride is used as anti-cancer potential for novel anticancer drugs.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications
Melting Point | ∼240°C (decomposition) |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 3563996 |
Solubility Information | Soluble in water,methanol and dimethyl sulfoxide. |
Formula Weight | 158.63 |
Percent Purity | 98% |
Chemical Name or Material | p-Tolylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'