Learn More
Thermo Scientific™ Brilliance™ Listeria Selective Supplement
Brilliance Listeria Selective Supplement is added to Listeria Agar Base to isolate/differentiate Listeria spp. in food samples.
Brand: Thermo Scientific™ SR0227E
Additional Details : Weight : 0.15000kg
Description
Brilliance Listeria Selective Supplement is added to Brilliance Listeria Agar Base, Mfr. No. CM1080E and Mfr. No. CM1080B, a chromogenic medium for the isolation and differentiation of Listeria spp. in food samples.
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both man and animals. Some Listeria ivanovii strains also possess lecithinase activity and, although Listeria ivanovii are primarily pathogenic to animals, there are strains which have been shown to cause infection in humans. Brilliance Listeria Agar uses the chromogen X-glucoside for presumptive identification of Listeria spp. This chromogen is cleaved by β-glucosidase which is common to all Listeria species. Other organisms that possess this enzyme, such as enterococci, are inhibited by the selective agents within the medium; lithium chloride, polymyxin B and nalidixic acid, while amphotericin inhibits the growth of any yeasts and moulds present in the sample. Listeria monocytogenes and pathogenic Listeria ivanovii are then further differentiated by their ability to produce the phospholipase enzyme, lecithinase. This enzyme hydrolyses the lecithin in the medium, producing an opaque white halo around the colony. Brilliance Listeria Agar is a modification of the formulation described by Ottaviani and Agosti. As in the original formulation, the medium is designed to identify Listeria spp. based on their utilization of a chromogenic substrate. However, in this modification, the pathogenic Listeria spp. are then further differentiated by the detection of lecithinase (phosphotidylcholine phospholipase C (PCPLC) activity, rather than phosphotidylinositol phospholipase C (PIPLC) activity. Both enzymes, PCPLC and PIPLC, are associated with virulence in Listeria spp., and, therefore, the presence of either enzyme is a useful indicator of pathogenicity.- Add to Brilliance Listeria Selective Agar Base, Part No. CM1080B
- Each vial supplements 500mL of medium
Specifications
Listeria | |
Freeze-dried | |
10/Pk. |
Brilliance Listeria Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Safety and Handling
- CHROMOGENIC LISTERIA Selective Supplement
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Supplemental information
- MIXTURE LIST-Contains : Nalidixic acid sodium salt, Polymyxin Bsulfate, Amphotericin B, Ceftazidime hydrate