Learn More
Thermo Scientific™ Brilliance™ Listeria Agar Base
Brilliance Listeria Agar Base, a chromogenic medium, isolates, enumerates and identifies Listeria spp. from food samples.
Brand: Thermo Scientific™ CM1080B
Additional Details : Weight : 0.50000kg
Description
Brilliance Listeria Agar Base is a chromogenic medium for the isolation, enumeration, and presumptive identification of Listeria spp., including Listeria monocytogenes.
Description
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both man and animals. Some Listeria ivanovii strains also possess lecithinase activity and, although Listeria ivanovii are primarily pathogenic to animals, there are strains which have been shown to cause infection in humans.
Brilliance Listeria Agar uses the chromogen X-glucoside for presumptive identification of Listeria spp. This chromogen is cleaved by β-glucosidase which is common to all Listeria species. Other organisms that possess this enzyme, such as enterococci, are inhibited by the selective agents within the medium; lithium chloride, polymyxin B and nalidixic acid, while amphotericin inhibits the growth of any yeasts and moulds present in the sample. Listeria monocytogenes and pathogenic Listeria ivanovii are then further differentiated by their ability to produce the phospholipase enzyme, lecithinase. This enzyme hydrolyses the lecithin in the medium, producing an opaque white halo around the colony.
Brilliance Listeria Agar is a modification of the formulation described by Ottaviani and Agosti. As in the original formulation, the medium is designed to identify Listeria spp. based on their utilization of a chromogenic substrate. However, in this modification, the pathogenic Listeria spp. are then further differentiated by the detection of lecithinase (phosphotidylcholine phospholipase C (PCPLC) activity, rather than phosphotidylinositol phospholipase C (PIPLC) activity. Both enzymes, PCPLC and PIPLC, are associated with virulence in Listeria spp., and, therefore, the presence of either enzyme is a useful indicator of pathogenicity. Also available Chromogenic Listeria Selective Supplement, Part No. SR0227E and Chromogenic Listeria Differential Supplement, Part No. SR0228E.
Appearance
- Dehydrated medium: Straw coloured, free-flowing powder
- Prepared medium: Translucent, white gel
Quality Control
Positive Controls - Expected Results:
- Listeria monocytogenes ATCC™ 7644™ - Good growth; blue/green colonies with haloes
- Listeria innocua ATCC™ 33090™ - Good growth; blue/green colonies with no halo
Negative Control - Expected Results:
- Enterococcus feacalis ATCC™ 29212™ - Inhibited
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and used under licence.
Specifications
Dehydrated medium should be stored at 10° to 30°C and used before the expiry date on the label. Prepared medium may be stored for up to 2 weeks at 2° to 8°C. | |
Powder | |
500 g |
Brilliance Listeria Agar Base | |
Agar | |
For 7.4L medium |
Safety and Handling
- Brilliance Listeria Agar Base
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Lithium chloride
ONE Broth-Listeria Base and ONE Broth Selective Supplement are for Laboratory Use Only.
ONE Broth-Listeria Base is designated as “Irritant” and ONE Broth-Listeria Selective Supplement is designated as “Harmful.” For a full Material Safety Data Sheet (MSDS), contact your Fisher Scientific Customer Service Representative.
Do not use ONE Broth-Listeria Base or ONE Broth-Listeria Selective Supplement beyond the stated expiration dates, or if the products show any sign of deterioration.