Learn More
Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers
Choose from two sizes for manual sequencing with Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers.
456.00 NOK
Specifications
For Use With (Equipment) | Owl™ S3S Aluminum-Backed Sequencer System |
---|---|
Product Type | Spacers |
Width (Metric) | 1.0 cm |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12474727
|
Thermo Scientific™
S2SL-SA4 |
45 cm |
456.00 NOK
Pack of 3 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Choose from two sizes for manual sequencing with Thermo Scientific™ Owl™ Spacer Sets for Aluminum-Backed Sequencers.
Available in 1.0W x 45cmL (0.4mm thick) or 1.0W x 65cmL (0.2mm thick).Specifications
Owl™ S3S Aluminum-Backed Sequencer System | |
Spacers | |
1.0 cm |