missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
362.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 1 g |
Solubility Information | Sparingly Soluble (0.61g/L) (25°C),Calc. |
Formula Weight | 1130.18 |
Physical Form | Powder |
Chemical Name or Material | Octakis (trimethylsiloxy)silsesquioxane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'