Learn More
Motic™ Objective, Plan Apochromat ELWD, Parfocality Adj. for PSM-1000
Extra-long working distance, parfocality adjustable objective suitable for PSM1000 Microscopes. Motic™ Objective, Plan Apochromat ELWD, Parfocality Adj. for PSM-1000 is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001700122
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15445612
Quantity Price
1 15867.00 NOK / Each
3 14756.31 NOK / Each
Estimated Shipment
15-01-2021
Add to basket
Description
Description
  • Plan apochromat objective.
  • Extra-long working distance.
  • Parfocality adjustable.
  • Spherical aberration correction: 3-4 colors.
  • Chromatic aberration correction: 4-5 colors.
  • Field curvature correction.
Specifications
Specifications
PSM-1000 Probe Station Microscope
Plan apochromat, extra-long working distance, parfocality adjustable
20X
WD: 20mm, NA: 0.420
Objective