Learn More
o-Xylene-d4 (ring-d4), CDN
Product Code. 30310324
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310324
Brand: CDN CDND39040.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material o-Xylene ring-d4
CAS 62367-40-2
Molecular Formula C8 2H4 H6
Quantity 0.5 g
Synonym o-Xylene D4 (Ring D4), 1,2,3,4-Tetradeuterio-5,6-dimethylbenzene, Benzene-1,2,3,4-d4, 5,6-dimethyl- (9 CI)
InChI Formula InChI=1 S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6 H,1-2H3/i3D,4 D,5 D,6 D
SMILES [2 H]c1c([2 H])c([2 H])c(C)c(C)c1[2 H]
IUPAC Name 1,2,3,4-tetradeuterio-5,6-dimethylbenzene
Molecular Weight (g/mol) 110.19
Formula Weight 110.1034 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity