Learn More
o-Toluic acid, 98+%, Thermo Scientific Chemicals
Catalog Number p-7021498
264.01 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1000 g
5000 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
118-90-1
C8H8O2
136.15
MFCD00002477
ZWLPBLYKEWSWPD-UHFFFAOYSA-N
o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
8373
CHEBI:36632
2-methylbenzoic acid
CC1=CC=CC=C1C(=O)O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
118-90-1
C8H8O2
136.15
MFCD00002477
ZWLPBLYKEWSWPD-UHFFFAOYSA-N
o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
8373
CHEBI:36632
2-methylbenzoic acid
CC1=CC=CC=C1C(=O)O
o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.
Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 103°C to 106°C
Density 1.06
Boiling Point 258°C to 259°C
Flash Point 148°C (298°F)
Odor Aromatic
Quantity 250 g
Beilstein 1072103
Merck Index 14,9535
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material o-Toluic acid