missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzylhydroxylamine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.
Solubility
Soluble in dimethylsulfoxide and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.
Solubility
Soluble in dimethylsulfoxide and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.058 |
Boiling Point | 118°C to 119°C (30 mmHg) |
Linear Formula | C6H5CH2ONH2 |
Refractive Index | 1.541 |
Quantity | 5 g |
UN Number | UN2735 |
Beilstein | 1906691 |
Solubility Information | Soluble in dimethylsulfoxide and methanol. |
Formula Weight | 123.15 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'