Learn More
Nur77 Monoclonal Antibody (12.14), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 53-5965-80
913.37 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.09500kg
Description
Description: This 12.14 monoclonal antibody reacts with mouse Nur77 (also known as NR4A1, TR3, NGFI-B, or NAK1), an inducible orphan nuclear receptor. Expressed in thymocytes and T cell lines, Nur77 promotes apoptosis and plays a role in thymocyte negative selection. Additionally, Nur77 has been shown to be critical for steroid biosynthesis in Leydig cells as well as for the effects of dopamine. In addition, Nur77 has been shown to interact with FoxP3 in regulatory T cells. However, our results with this antibody do not correlate with this observation. Applications Reported: This 12.14 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 12.14 antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Nur77 Monoclonal antibody specifically detects Nur77 in Mouse samples. It is validated for Flow Cytometry.Specifications
Nur77 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P12813 | |
NR4A1 | |
Affinity chromatography | |
RUO | |
15370 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
12.14 | |
Alexa Fluor 488 | |
NR4A1 | |
early response protein NAK1; Gfrp; GFRP1; growth factor-inducible nuclear protein N10; Hbr1; Hbr-1; Hmr; hormone receptor; immediate early gene transcription factor NGFI-B; MGC9485; N10; NAK1; NAK-1; nerve growth factor IB nuclear receptor variant 1; nerve growth factor induced protein I-B; nerve growth factor-induced protein I-B; Ngfib; NGFI-B; NP10; Nr4a1; Nuclear hormone receptor NUR/77; nuclear protein N10; Nuclear receptor subfamily 4 group A member 1; nuclear receptor subfamily 4, group A, member 1; NUR77; Orphan nuclear receptor HMR; orphan nuclear receptor TR3; Orphan receptor tr3; ST-59; steroid receptor TR3; testicular receptor 3; TIS1; TR3; TR3 orphan receptor | |
Mouse | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.