missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NUP98 Recombinant Protein

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH12906.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 4928
Molecular Weight (g/mol) 92.4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16188531
View Documents Promotion Details
Abnova™
H00004928-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16198531
View Documents Promotion Details
Abnova™
H00004928-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH12906.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
92.4
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NUP98
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
4928
NUP98 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLYQTPLELKLKHSTVHVDELCPLIVPNLGVAVIHDYADWVKEASGDLPEAQIVKHWSLTWTLCEALWGHLKELDSQLNEPREYIQILERRRAFSRWLSCTATPQIEEEVSLTQKNSPVEAVFSYLTGKRISEACSLAQQSGDHRLALLLSQFVGSQSVRELLTMQLVDWHQLQADSFIQDERLRIFALLAGKPVWQLSEKKQINVCSQLDWKRSLAIHLWYLLPPTASISRALSMYEEAFQNTSDSDRYACSPLPSYLEGSGCVIAEEQNSQTPLRDVCFHLLKLYSDRHYDLNQLLEPRSITADPLDYRLSWHLWEVLRALNYTHLSAQCEGVLQASYAGQLESEGLWEWAIFVLLHIDNSGIREKAVRELLTRHCQLLETPESWAKETFLTQKLRVPAKWIHEAKAVRAHMESDKHLEALCLFKAEHWNRCHKLIIRHLASDAIINENYDYLKGFLEDLAPPERSSLIQDWETSGLVYLDYIRVIEMLRHIQQVDCSGNDLEQLHIKVTSLCSRIEQIQCYSAKDRLAQSDMAKRVANLLRVVLSLHHPPDRTSDSTPDPQRVPLRLLAPHIGRLPMPEDYAMDELRSLTQSYLRELAVGSL
ADIR2/NUP196/NUP96
NUP98
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications