Learn More
Macherey-Nagel™ Nucleodur™ SiOH HPLC Columns
Column Format: Analytical Column; Pore Size: 110 Å; Column Phase: Normal Phase
3135.00 NOK - 5230.00 NOK
Specifications
Column Chemistry | High purity Silica |
---|---|
Column Format | Analytical Column |
Crosslinking | High purity Silica |
Diameter (Metric) | 4.6 mm |
For Use With | For Analytical and Preparative Separation of Polar and Midpolar Compounds Applications |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12498377
|
Macherey-Nagel™
760172.46 |
150 mm |
4295.00 NOK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12727810
|
Macherey-Nagel™
760173.46 |
250 mm |
5230.00 NOK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
12717810
|
Macherey-Nagel™
760170.46 |
50 mm |
3135.00 NOK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Specifications
High purity Silica | |
High purity Silica | |
For Analytical and Preparative Separation of Polar and Midpolar Compounds Applications | |
8700 psi (600 bar) | |
n-heptane | |
3 μm | |
110 Å | |
1 Ea. | |
340 m²/g | |
L3 | |
Silica | |
No | |
Stainless Steel | |
Analytical HPLC |
Analytical Column | |
4.6 mm | |
Polar and midpolar | |
<10 ppm | |
Spherical | |
2 to 8 | |
0.9 mL/g | |
n-heptane | |
60°C | |
Silica | |
Normal Phase | |
Normal Phase | |
EC Column |