Learn More
Merck Millipore Novagen™ KOD Hot Start Master Mix
For convenient high fidelity PCR
Brand:  Merck Millipore 71842-3
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15448911
  • 3406.00 NOK / 100 reactions
Estimated Shipment
12-01-2021
Add to basket
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
Specifications
Specifications
100 rxn
-20°C
Videos
SDS
Documents
For Research Use Only