Learn More
Merck Millipore Novagen™ KOD Hot Start Master Mix
For convenient high fidelity PCR
Brand: Merck Millipore 71842-4
Additional Details : Weight : 0.05000kg
Description
- High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
- Amplifies genomic DNA templates up to 12 kbp
- Amplifies plasmid and lambda DNA template up to 21 kbp
- Eliminates mispriming and primer-dimer formation
- Convenient ambient-temperature setup compatible with automation
- KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
Specifications
500 rxn | |
-20°C |
For Research Use Only