missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 65% w/w, Pharmpur™, Scharlab

Catalog Number p-7083580
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

Suitable for determinations with dithizone. Scharlab Nitric Acid, 65% w/w, Pharmpur™ is also used as an oxidizing agent, for the synthesis of nitrates and organic nitro compounds, and in the pharmaceutical industry.

  • Molecular Formula: HNO3
  • Molar Mass: 63.01g/mol
  • CAS: 7697-37-2
  • EIN ECS No.: 231-714-2
Melting Point ∼-32°C
Boiling Point 122°C
Density 1.41g/cm3
Residue after Ignition 0.0003% max
Vapor Pressure 9.4hPa at 20°C
Quantity 1 L
Physical Form Liquid
Solubility Information Miscible in water (20°C)
Formula Weight 63.01g/mol
Percent Purity ≥65% (acidimetric)
Grade Pharmpur™
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitric acid, solution min. 65% w/w, Pharmpur, ISO, Reag. Ph Eur, for determinations with dithizone
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'