missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nile red, MP Biomedicals™
201.89 NOK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
10 mg
250 mg
500 mg
Specifications
Formula Weight | 318.4 |
Color | Green |
Quantity | 10 mg |
Physical Form | Powder |
Chemical Name or Material | Nile red, Determination of lipids, staining whole tissue |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'