Learn More
R&D Systems NHP XL CCL2/MCP-1, Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | CCL2/JE/MCP-1 |
| Gene ID (Entrez) | 6347 |
| Synonym | C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |