Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
For C76 Water Bath Shaker
Brand: Eppendorf™ M1231-2000
Additional Details : Weight : 0.80000kg
Description
- Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
Specifications
Model C76 water bath shaker | |
Stainless Steel |