Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
For C76 Water Bath Shaker
Brand:  Eppendorf™ M1231-2000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11389874
  • 7715.00 NOK / Each
Estimated Shipment
26-01-2021
Add to basket
Description
Description
  • Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
Specifications
Specifications
Model C76 water bath shaker