missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%

Brand:  Alfa Aesar™ H63331.03

Additional Details : CAS Number : 147290-11-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429978

  • 338.27 NOK / 1g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

147290-11-7
438.48
RXLIOYNXBHZZBI-NRFANRHFSA-N
2756120
C=CCOC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C24H26N2O6
MFCD00798637
fmoc-orn alloc-oh, a-alloc-l-ornithine, n-a-boc-n-d-allyloxycarbonyl-l-ornithine, n-alpha-9-fluorenylmethyloxycarbonyl-n-delta-allyloxycarbonyl-l-ornithine, fmoc-orn aloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy carbonyl amino pentanoic acid, n2-fmoc-n5-allyloxycarbonyl-l-ornithine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-prop-2-en-1-yloxy carbonyl amino pentanoic acid, pubchem6308, n, a-alloc-n, a-fmoc-l-ornithine, a-fmoc-n
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-(prop-2-enoxycarbonylamino)pentanoic acid

Specifications

Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine
C24H26N2O6
fmoc-orn alloc-oh, a-alloc-l-ornithine, n-a-boc-n-d-allyloxycarbonyl-l-ornithine, n-alpha-9-fluorenylmethyloxycarbonyl-n-delta-allyloxycarbonyl-l-ornithine, fmoc-orn aloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy carbonyl amino pentanoic acid, n2-fmoc-n5-allyloxycarbonyl-l-ornithine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-prop-2-en-1-yloxy carbonyl amino pentanoic acid, pubchem6308, n, a-alloc-n, a-fmoc-l-ornithine, a-fmoc-n
C=CCOC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
438.48
438.47
95%
147290-11-7
MFCD00798637
RXLIOYNXBHZZBI-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-(prop-2-enoxycarbonylamino)pentanoic acid
2756120
Solid
1g
Videos
SDS Documents