Learn More
Thermo Scientific™ National Storerooms with Target™ Crimp-Top Vials
Your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
Brand: Thermo Scientific™ C4011-688
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ National™ Storerooms with Target Crimp-Top Vials feature your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
- Mini Storeroom has 6 drawers; each drawer contains 100 packs of vials or septa
- Full size storeroom has 9 deep drawers with a capacity of 2000 vials and seals
- Stackable cabinet constructed of heavy-gauge steel
- With metal shelves and injection-molded drawers
- Storeroom can be restocked as product is consumed
Includes:Prestocked with 3000 each of: clear vials and preassembled clear crimp seals with FEP/red rubber septa
Specifications
Clear | |
Clear | |
PTFE/RR Seals |
NSC Mini Storeroom, Clear Vial, PTFE/RR Seals | |
PTFE | |
Crimp Cap |