Learn More
Thermo Scientific™ National Storerooms with Target™ Crimp-Top Vials
Your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
Brand:  Thermo Scientific™ C4075-21
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12470308
Quantity Price
1 7193.00 NOK / Each
5 6690.93 NOK / Each
Estimated Shipment: 09-11-2021
Add to basket
Description
Description
Thermo Scientific™ National™ Storerooms with Target Crimp-Top Vials feature your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
  • Mini Storeroom has 6 drawers; each drawer contains 100 packs of vials or septa
  • Full size storeroom has 9 deep drawers with a capacity of 2000 vials and seals
  • Stackable cabinet constructed of heavy-gauge steel
  • With metal shelves and injection-molded drawers
  • Storeroom can be restocked as product is consumed
Includes:Prestocked with 3000 each of: clear vials and preassembled clear crimp seals with FEP/red rubber septa
Specifications
Specifications
Clear
Clear
PTFE/RR Seals
Yes
NSC Storeroom, Clear I-D Vial
PTFE
Crimp