Learn More
Alfa Aesar™ Nalpha-Fmoc-N-gamma-trityl-D-asparagine, 97%
Brand:  Alfa Aesar™ H63194.03
Additional Details : CAS Number : 180570-71-2 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15429758
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
180570-71-2
596.683
KJYAFJQCGPUXJY-UUWRZZSWSA-N
24820182
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
C38H32N2O5
MFCD00151919
fmoc-d-asn trt-oh, n-fmoc-n'-trityl-d-asparagine, nalpha-fmoc-ngamma-trityl-d-asparagine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethylcarbamoyl propanoic acid, pubchem12398, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-oxo-4-tritylamino butanoic acid, n~2~-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-asparagine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-oxidanylidene-4-triphenylmethyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy oxo methyl amino-4-oxo-4-triphenylmethyl amino butanoic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanoic acid
Specifications
Nalpha-Fmoc-N-gamma-trityl-D-asparagine
C38H32N2O5
fmoc-d-asn trt-oh, n-fmoc-n'-trityl-d-asparagine, nalpha-fmoc-ngamma-trityl-d-asparagine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethylcarbamoyl propanoic acid, pubchem12398, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-oxo-4-tritylamino butanoic acid, n~2~-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-asparagine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-oxidanylidene-4-triphenylmethyl amino butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy oxo methyl amino-4-oxo-4-triphenylmethyl amino butanoic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
596.683
596.67
97%
180570-71-2
MFCD00151919
KJYAFJQCGPUXJY-UUWRZZSWSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-(tritylamino)butanoic acid
24820182
Solid
1g