Learn More
Nalidixic Acid Methyl Ester, Mikromol
Product Code. 30459720
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30459720
Brand: Mikromol MM3144.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nalidixic Acid Methyl Ester, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nalidixic Acid Methyl Ester
CAS 63475-29-6
Analyte or Component Names Nalidixic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C13 H14 N2 O3
Certifications/Compliance ISO 9001
Synonym 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid methyl ester, Methyl nalidixate, Nalidixic acid methyl ester
InChI Formula InChI=1S/C13H14N2O3/c1-4-15-7-10(13(17)18-3)11(16)9-6-5-8(2)14-12(9)15/h5-7H,4H2,1-3H3
Solution Type Neat
SMILES CCN1C=C(C(=O)OC)C(=O)c2ccc(C)nc12
IUPAC Name methyl 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
Molecular Weight (g/mol) 262.27
Quantity 4 x 25 mg
Formula Weight 262.265 g/mol