Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with SFCA Membrane
Lower the number of extractables in your bottle-top filters by switching from standard CA membranes to sterile, disposable bottle top filters with SFCA Membrane.
755.00 NOK - 1720.00 NOK
Specifications
No. per Case 12
No. per Pack 12
Sterility Sterile
View More Specs
Products 10
Product Code Brand Volume (Metric) Pore Size (Metric) Fits Bottle Neck Size (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Pore Size (Metric) Fits Bottle Neck Size (Metric) Price Quantity & Availability  
10352911 View Documents Thermo Scientific™
290-3320
150 mL 0.2 μm 33 mm 755.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
07-05-2020
Add to basket
10027780 View Documents Thermo Scientific™
290-3345
150 mL 0.45 μm 33 mm 755.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
07-05-2020
Add to basket
10139560 View Documents Thermo Scientific™
290-4520
150 mL 0.2 μm 45 mm 755.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
29-04-2020
Add to basket
10624281 View Documents Thermo Scientific™
290-4545
150 mL 0.45 μm 45 mm 755.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
29-04-2020
Add to basket
10037790 View Documents Thermo Scientific™
291-3320
500 mL 0.2 μm 33 mm 1060.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
07-05-2020
Add to basket
10529051 View Documents Thermo Scientific™
291-3345
500 mL 0.45 μm 33 mm 1060.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Call for latest availability
+47 22 95 59 59
Add to basket
10201371 View Documents Thermo Scientific™
291-4520
500 mL 0.2 μm 45 mm 1059.05 NOK
Pack of 12 View alternative products
Save Up to 
»
In Stock 8
Add to basket
10549241 View Documents Thermo Scientific™
291-4545
500 mL 0.45 μm 45 mm 1060.00 NOK
Pack of 12 View alternative products
Save Up to 
»
In Stock 5
Add to basket
10139800 View Documents Thermo Scientific™
292-3320
1000 mL 0.2 μm 33 mm 1720.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
07-05-2020
Add to basket
10790592 View Documents Thermo Scientific™
292-4520
1000 mL 0.2 μm 45 mm 1720.00 NOK
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
07-05-2020
Add to basket
Description
Description
Bottle Top Filters:
  • Marked with catalog number, membrane type, pore size, lot number and expiration date for easy traceability
  • Gamma radiation sterilized and individually bagged for 5-year sterile shelf life
  • Molded-in graduations for quick visualization of sample volume
  • Side-arm with quick-disconnect tubing adapter for easy bottle top changes
  • Non-cytotoxic and non-pyrogenic
Surfactant-Free Cellulose Acetate Membranes (SFCA)
  • Low protein binding with little or no loss of specific proteins
  • Excellent flow rates
  • No wetting agents to affect sensitive cell culture lines
  • Much lower extractables than standard CA membranes 
Includes:
Polystyrene graduated chamber; color-coded collar with cellulosic vent plug side arm and quick-disconnect tubing adapter; 50, 75, or 90mm diameter membranes; bottom screw threads for attachment to bottles with 33 or 45mm neck sizes.
Warranty: 90 days
Specifications
Specifications
12
12
Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.