missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ 4mm Syringe Filters
Filter sample volume sizes of 0.5 to 1.0mL using Nalgene 4mm Syringe Filters.
1710.00 NOK
Product Code | Brand | Membrane | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | Pore Size | Price | Quantity & Availability | ||||
10303201 |
Thermo Scientific™
171-0020 |
Cellulose Acetate | 0.2μm |
1710.00 NOK Pack of 100 |
|
10538861 |
Thermo Scientific™
171-0045 |
Cellulose Acetate | 0.45μm |
1710.00 NOK Pack of 100 |
|
10353291 |
Thermo Scientific™
176-0020 |
Nylon | 0.2μm |
1710.00 NOK Pack of 100 |
|
10722732 |
Thermo Scientific™
176-0045 |
Nylon | 0.45μm |
1710.00 NOK Pack of 100 |
|
Description
Cellulose Acetate (CA)
- Low protein binding
- Ideal for filtration of biological/immunological samples, cell culture media components, biological fluids, most other aqueous solutions and reagents, including EIA and ELISA samples
Nylon
- Naturally hydrophilic, low extractables
- Useful for filtering cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC samples and solvents
Specifications
4mm | |
400 | |
100 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.