Learn More
Naepaine Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Naepaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Naepaine Hydrochloride |
| CAS | 614-42-6 |
| Analyte or Component Names | Naepaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 N2 O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethanol, 2-(pentylamino)-, 1-(4-aminobenzoate), hydrochloride (1:1), Ethanol, 2-(pentylamino)-, 4-aminobenzoate (ester), monohydrochloride (9CI), Ethanol, 2-(pentylamino)-, p-aminobenzoate (ester), monohydrochloride (8CI), Amylsine hydrochloride, Naepaine hydrochloride |
| InChI Formula | InChI=1S/C14H22N2O2.ClH/c1-2-3-4-9-16-10-11-18-14(17)12-5-7-13(15)8-6-12;/h5-8,16H,2-4,9-11,15H2,1H3;1H |
| SMILES | Cl.CCCCCNCCOC(=O)c1ccc(N)cc1 |
| IUPAC Name | 2-(pentylamino)ethyl 4-aminobenzoate;hydrochloride |
| Show More |