Learn More
n-Triacontane, 98+%
Descrizione
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.
Solubility
Soluble in benzene, chloroform and toluene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifica
Specifica
| Melting Point | 65°C to 68°C |
| Boiling Point | 455°C |
| Flash Point | 237°C (458°F) |
| Odor | Odorless |
| Quantity | 1 g |
| Beilstein | 1777281 |
| Solubility Information | Soluble in benzene,chloroform and toluene. Insoluble in water. |
| Formula Weight | 422.83 |
| Percent Purity | 99% |
| Chemical Name or Material | n-Triacontane |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.