missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 1-Fmoc-piperidine-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7039437
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
1823301-50-3
C25H24N2O6
448.475
MFCD22988969
RDBHMYCVGMHUQA-UHFFFAOYSA-N
4-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl piperidine-1,4-dicarboxylate, succinimidyl 1-fmoc-piperidine-4-carboxylate
98043111
4-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) piperidine-1,4-dicarboxylate
C1CN(CCC1C(=O)ON2C(=O)CCC2=O)C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
This item is not returnable. View return policy
1823301-50-3
C25H24N2O6
448.475
MFCD22988969
RDBHMYCVGMHUQA-UHFFFAOYSA-N
4-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl piperidine-1,4-dicarboxylate, succinimidyl 1-fmoc-piperidine-4-carboxylate
98043111
4-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) piperidine-1,4-dicarboxylate
C1CN(CCC1C(=O)ON2C(=O)CCC2=O)C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 2 g
Formula Weight 448.48
Percent Purity 97%
Chemical Name or Material N-Succinimidyl 1-Fmoc-piperidine-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'