missing translation for 'onlineSavingsMsg'
Learn More

n-Nonane, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-3737088
Click to view available options
Quantity:
2 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-84-2
MFCD00009574
BKIMMITUMNQMOS-UHFFFAOYSA-N
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
8141
CHEBI:32892
nonane
CCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
111-84-2
MFCD00009574
BKIMMITUMNQMOS-UHFFFAOYSA-N
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
8141
CHEBI:32892
nonane
CCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7180g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H20
Linear Formula CH3(CH2)7CH3
Refractive Index 1.4040 to 1.4060 (20°C, 589nm)
Quantity 2 mL
Beilstein 01, 165
Molecular Weight (g/mol) 128.259
Formula Weight 128.26
Percent Purity ≥98.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Nonane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'