missing translation for 'onlineSavingsMsg'
Learn More

N,N',N''-Triphenylguanidine, 97%, Thermo Scientific™

Catalog Number p-7024336
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
101-01-9
C19H17N3
287.366
MFCD00020648
FUPAJKKAHDLPAZ-UHFFFAOYSA-N
n,n',n-triphenylguanidine, guanidine, n,n',n-triphenyl, sym-triphenylguanidine, guanidine, triphenyl, 1,2,3-trifenylguanidin, unii-i64b170qfg, 1,2,3-trifenylguanidin czech, guanidine, 1,2,3-triphenyl, phenyl 2-phenyl-1-phenylamino-2-azavinyl amine, triphenylguanidin
7539
1,2,3-triphenylguanidine
C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3
This item is not returnable. View return policy
101-01-9
C19H17N3
287.366
MFCD00020648
FUPAJKKAHDLPAZ-UHFFFAOYSA-N
n,n',n-triphenylguanidine, guanidine, n,n',n-triphenyl, sym-triphenylguanidine, guanidine, triphenyl, 1,2,3-trifenylguanidin, unii-i64b170qfg, 1,2,3-trifenylguanidin czech, guanidine, 1,2,3-triphenyl, phenyl 2-phenyl-1-phenylamino-2-azavinyl amine, triphenylguanidin
7539
1,2,3-triphenylguanidine
C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 142°C to 145°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 287.36
Percent Purity 97%
Chemical Name or Material N,N',N''-Triphenylguanidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'